Learn More
Aviva Systems Biology Corp BFSP2 Antibody - middle region (ARP64902_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64902_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BFSP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human BFSP2. Alternate names: CP47, CP49, LIFL-L, MGC142078, MGC142080, PHAKOSINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-388