Learn More
Siemens Healthineers BFT II Analyzer
Catalog No. 23044714 Shop All Siemens Healthineers Products
Change view
Click to view available options
Throughput:
60 PT Tests/Hr., 30 APTT Tests/Hr.
Catalog No. Throughput
23-044-714 60 PT Tests/Hr., 30 APTT Tests/Hr.
1 options
This item is not returnable. View return policy
Catalog No. 23-044-714 Supplier Siemens Healthineers Supplier No. 10458677
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
This item is not returnable. View return policy
The BFT II Analyzer is a semi-automated device intended for use to determine PT, APTT and Fibrinogen.
The BFT II Analyzer combines the advantages of mechanical and photo-optical clot-detecting principles by employing a unique turbodensitometric detection device. The semi-automated, dual-channel coagulometer is ideal for low-volume laboratories performing routine coagulation tests.
  • Light protection cap reduces the risk of user errors by keeping the pipetting channel closed unless a cuvette has been inserted.
  • Pre-defined assays with stored reference curves.
  • Automatic calculation of PT in sec, % of norm, INR, Fibrinogen in mg/dL or g/L.
  • Dual measuring channels allow for simultaneous determinations.
  • Half-volume and single cuvette testing require minimal sample volume.
  • Built-in thermal printer for consolidated result handling.
TRUSTED_SUSTAINABILITY
Specifications
Type BFT II Analyzer
Throughput 60 PT Tests/Hr., 30 APTT Tests/Hr.
Cuvettes 30 Cuvettes
Depth (Metric) 300 mm
Height (Metric) 100 mm
Width (Metric) 200 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov