Learn More
New England Biolabs, Inc. BfuAI – 1250 units
SDP
Catalog No. 50812407
Encompass
A restriction endonuclease that recognizes the sequence ACCTGCNNNN^NNNN_.
Catalog No. 50-812-407 Supplier New England Biolabs, Inc. Supplier No. R0701L
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
A restriction endonuclease that recognizes the sequence ACCTGCNNNN^NNNN_.