Learn More
New England Biolabs, Inc. BfuAI – 250 units
SDP
Catalog No. 50812408
Encompass
A restriction endonuclease that recognizes the sequence ACCTGCNNNN^NNNN_.
Catalog No. 50-812-408 Supplier New England Biolabs, Inc. Supplier No. R0701S
A restriction endonuclease that recognizes the sequence ACCTGCNNNN^NNNN_.