Learn More
Bioworld BG-A1 Medium, 25 L
Supplier:  Bioworld 306302092
Encompass_Preferred
For infection and co-cultivation of stolon nodes with Agrobacterium tumefaciens for production of transgenic Bermuda grass. false false false Specification:Appearance: Cream to yellow powderSolublity: Soluble in waterRecommended use: 2.6g/LContains: Half strength Murashige & Skoog medium supplemented with 4.5uM Kinetin, 1.uM 2,4-D, 3.3 mM L-Cysteine.Add: Aseptically filter sterilized DTT (1mM), sodium thisulfate (1mM) and sucrose (2%) to the pre-sterilized BG-A1 medium or as required.
Catalog No. 50-255-024