Learn More
Bioworld BG-A2 Medium, 25 L
Catalog No. 50255027
Encompass_Preferred
For selection and recovery of transgenic plants (eg., Bermuda grass) infected with Agrobacterium tumefaciens.Specification:Appearance: cream to yellow powderSolubility: Partially soluble in waterRecommenced use: 12.4g/LContains: Murashige & Skoog medium supplemented with 4.5uM Kinetin, 0.2uM 2,4-D, 0.8% agar.Add: Aseptically add filter-sterilized hygromycin (75mg/L), silver nitrate (1.7mg/L), cefotaxime (250,g/L) and sucrose (2%) to the pre-sterilized BG-A2 medium or as required.
Catalog No. 50-255-027 Supplier Bioworld Supplier No. 306302102
For selection and recovery of transgenic plants (eg., Bermuda grass) infected with Agrobacterium tumefaciens.Specification:Appearance: cream to yellow powderSolubility: Partially soluble in waterRecommenced use: 12.4g/LContains: Murashige & Skoog medium supplemented with 4.5uM Kinetin, 0.2uM 2,4-D, 0.8% agar.Add: Aseptically add filter-sterilized hygromycin (75mg/L), silver nitrate (1.7mg/L), cefotaxime (250,g/L) and sucrose (2%) to the pre-sterilized BG-A2 medium or as required.