Learn More
Foxx Life Sciences EZBio Ultra Bag Assembly, Single Use BioProcess, 2D, 5L, PE Bag, 2x 3/8" port with Plug, 1x1/4"HB with needleless Luer insert, EZBio TPE Tubing, Sterile, 10/CS
Catalog No. 502886456
Encompass_Preferred
Foxx Life Sciences offers its high-performance Single-Use Bag Assembly for efficient fluid storage and transfer in critical bioprocessing. Designed for optimized flow with minimal fluid hold-up, these bags ensure reliable, high-throughput performance. Made from high-clarity medical-grade Renolit 9101 PE multilayer film, they meet ISO & USP biocompatibility standards, offering low oxygen/CO₂ permeability and excellent low-temperature performance.
Available in sizes 50mL–50L with edge ports and 20L and 50L with face ports, customizable up to 4 edge ports, double-bagged, and gamma-sterilized in the U.S.
Features
Optimized ports for enhanced flow rates & integrity
Customizable MTO assemblies with multiple port configurations
Low-profile design minimizes product hold-up
Applications
Buffer and media storage
Bulk product collection and transport
Chromatography and filtration buffer management
Fraction collection and product sampling
Catalog No. 50-288-6456 Supplier Foxx Life Sciences Supplier No. 66P600117FLS
Foxx Life Sciences offers its high-performance Single-Use Bag Assembly for efficient fluid storage and transfer in critical bioprocessing. Designed for optimized flow with minimal fluid hold-up, these bags ensure reliable, high-throughput performance. Made from high-clarity medical-grade Renolit 9101 PE multilayer film, they meet ISO & USP biocompatibility standards, offering low oxygen/CO₂ permeability and excellent low-temperature performance.
Available in sizes 50mL–50L with edge ports and 20L and 50L with face ports, customizable up to 4 edge ports, double-bagged, and gamma-sterilized in the U.S.
Features
Optimized ports for enhanced flow rates & integrity
Customizable MTO assemblies with multiple port configurations
Low-profile design minimizes product hold-up
Applications
Buffer and media storage
Bulk product collection and transport
Chromatography and filtration buffer management
Fraction collection and product sampling