Learn More
New England Biolabs, Inc. BG-GLA-NHS – 2 mg
SDP
Catalog No. 508112076
Encompass
BG-GLA-NHS is an amine-reactive building block for the one-step synthesis of SNAP-tag substrates from amine-containing precursors including proteins, peptides or oligonucleotides.
Catalog No. 50-811-2076 Supplier New England Biolabs, Inc. Supplier No. S9151S
BG-GLA-NHS is an amine-reactive building block for the one-step synthesis of SNAP-tag substrates from amine-containing precursors including proteins, peptides or oligonucleotides.