Learn More
New England Biolabs, Inc. BG-Maleimide – 2 mg
SDP
Catalog No. 508112077
Encompass
BG-Maleimide is a thiol-reactive building block for the one-step synthesis of SNAP-tag substrates from thiol-containing precursors such as thiol-modified oligonucleotides.
Catalog No. 50-811-2077 Supplier New England Biolabs, Inc. Supplier No. S9153S
Add to Cart
Add to Cart
BG-Maleimide is a thiol-reactive building block for the one-step synthesis of SNAP-tag substrates from thiol-containing precursors such as thiol-modified oligonucleotides.