Learn More
MSC B&G Equipment Sprayer, 1 Gallon, 18 in Extension, Extenda-Ban Valve, 4-Way Tip
Catalog No. 502839284
Encompass
The Primeline Sprayer is a highly reliable and durable solution for professional pest control applications. Designed with high-quality stainless steel, special grade brass, and chemical-resistant gaskets and hoses, it ensures extended use and consistent performance when applying water or oil-based insecticides and disinfectants.
  • Stainless steel tank for durability
  • Durable brass grip and wand
  • Premium rubber hose with metal spring strain relief to prevent kinks
  • Extenda-Ban valve for instant tip shut-off
  • 4-way tip for versatile spray patterns (fine fan, coarse fan, pin stream)
  • Integrated tank holster for wand storage
Catalog No. 50-283-9284 Supplier MSC Supplier No. 74334400
The Primeline Sprayer is a highly reliable and durable solution for professional pest control applications. Designed with high-quality stainless steel, special grade brass, and chemical-resistant gaskets and hoses, it ensures extended use and consistent performance when applying water or oil-based insecticides and disinfectants.
  • Stainless steel tank for durability
  • Durable brass grip and wand
  • Premium rubber hose with metal spring strain relief to prevent kinks
  • Extenda-Ban valve for instant tip shut-off
  • 4-way tip for versatile spray patterns (fine fan, coarse fan, pin stream)
  • Integrated tank holster for wand storage