Learn More
New England Biolabs, Inc. BG-PEG-NH2 – 2 mg
SDP
Catalog No. 508112075
Encompass
BG-PEG-NH2 is an amine-terminated building block for the one-step synthesis of SNAP-tag substrates from NHS esters or other activated carboxyl esters on labels or surfaces.
Catalog No. 50-811-2075 Supplier New England Biolabs, Inc. Supplier No. S9150S
Add to Cart
Add to Cart
BG-PEG-NH2 is an amine-terminated building block for the one-step synthesis of SNAP-tag substrates from NHS esters or other activated carboxyl esters on labels or surfaces.