Learn More
IHEALTH LABS INC iHealth Smart Wireless Bluetooth Blood Glucose Meter
SDP
Catalog No. NC3451878
Encompass_Preferred
1. Seamless Compatibility- Our device is compatible with both iOS and Android, ensuring effortless integration with your phone/tablet.2. Proven Accuracy- Clinically tested and validated, you can rely on the accuracy of the readings it provides.3. Secure Data Storage- Your valuable health readings are safely saved on the secure iHealth cloud, giving you peace of mind.4. Easy Sharing- Share your readings with your doctor or family members directly from your phone/tablet, fostering better communication and collaboration in managing your health.Specifications- Model- BG5S Color- whiteProduct dimensions-3.85 inchesx1.37 inchesx1.09 inches (98mmx35mmx27.8mm)Product weight - 0.08lbs (0.035kg)Power source- DC 3.7V. Li-ion 250mAhCharging condition- DC 5V 250mAhBundle includes- 1 Health Smart Gluco-monitor, 1 Lancing Device, 10 Lancets, and a Travel Case.
This item is not returnable. View return policy
Catalog No. NC3451878 Supplier IHEALTH LABS INC Supplier No. BG5S
1. Seamless Compatibility- Our device is compatible with both iOS and Android, ensuring effortless integration with your phone/tablet.2. Proven Accuracy- Clinically tested and validated, you can rely on the accuracy of the readings it provides.3. Secure Data Storage- Your valuable health readings are safely saved on the secure iHealth cloud, giving you peace of mind.4. Easy Sharing- Share your readings with your doctor or family members directly from your phone/tablet, fostering better communication and collaboration in managing your health.Specifications- Model- BG5S Color- whiteProduct dimensions-3.85 inchesx1.37 inchesx1.09 inches (98mmx35mmx27.8mm)Product weight - 0.08lbs (0.035kg)Power source- DC 3.7V. Li-ion 250mAhCharging condition- DC 5V 250mAhBundle includes- 1 Health Smart Gluco-monitor, 1 Lancing Device, 10 Lancets, and a Travel Case.