Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BGG Stock Solution, acetylated

Catalog No. P2045
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2045 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2045 Supplier Invitrogen™ Supplier No. P2045
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The material is tested for level of background fluorescence.

Invitrogen's Acetylated BGG solution is a 0.5% solution in water.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mL
Content And Storage BGG stock solution is shipped on wet ice and should be stored at 4°C.
Shipping Condition Wet Ice
Protein Subtype BGG
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.