Learn More
Medchemexpress LLC HY-100600 5mg Medchemexpress, BGG463 CAS:890129-26-7 Purity:>98%
SDP
Catalog No. 501931897
Encompass_Preferred
Medchemexpress, HY-100600 5mg BGG463 CAS:890129-26-7 BGG463 (K03859) is a type II CDK2 inhibitor [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1897 Supplier Medchemexpress LLC Supplier No. HY1006005MG
Medchemexpress, HY-100600 5mg BGG463 CAS:890129-26-7 BGG463 (K03859) is a type II CDK2 inhibitor [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.