Learn More
Apexbio Technology LLC BGJ398(Synonyms: Infigratinib, NVP-BGJ398, BGJ-398, BGJ 398), 200mg, CAS: 872511-34-7.
SDP
Catalog No. 5000060769
Encompass_Preferred
BGJ398 (CAS 872511-34-7) also known as NVP-BGJ398 is an orally bioavailable small-molecule inhibitor targeting fibroblast growth factor receptors (FGFRs) It selectively inhibits receptor tyrosine kinase activity of FGFR1 FGFR2 FGFR3 and FGFR4 which are critical mediators in signal transduction pathways involved in cell proliferation differentiation and survival In preclinical studies BGJ398 suppresses proliferation and induces apoptosis in FGFR-dependent cancer cells additionally it demonstrates antitumor activity in xenograft tumor models BGJ398 is utilized as a research tool in oncology studies investigating FGFR-driven malignancies
Catalog No. 50-000-60769 Supplier Apexbio Technology LLC Supplier No. A3014200
BGJ398 (CAS 872511-34-7) also known as NVP-BGJ398 is an orally bioavailable small-molecule inhibitor targeting fibroblast growth factor receptors (FGFRs) It selectively inhibits receptor tyrosine kinase activity of FGFR1 FGFR2 FGFR3 and FGFR4 which are critical mediators in signal transduction pathways involved in cell proliferation differentiation and survival In preclinical studies BGJ398 suppresses proliferation and induces apoptosis in FGFR-dependent cancer cells additionally it demonstrates antitumor activity in xenograft tumor models BGJ398 is utilized as a research tool in oncology studies investigating FGFR-driven malignancies