Learn More
Selleck Chemical LLC Infigratinib (BGJ398) 5mg 872511-34-7 NVP-BGJ398
SDP
Supplier:  Selleck Chemical LLC S21835MG
Encompass_Preferred
Infigratinib (BGJ398) is a potent and selective FGFR inhibitor for FGFR1/2/3 with IC50 of 0.9 nM/1.4 nM/1 nM in cell-free assays, >40-fold selective for FGFR versus FGFR4 and VEGFR2, and little activity to Abl, Fyn, Kit, Lck, Lyn and Yes. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0382493