Learn More
Cayman Chemical BGJ398phosphate 10mg
SDP
Catalog No. 502917233
Encompass_Preferred
A pan FGFR inhibitor (IC50s = 0.9, 1.4, and 1 nM for FGFR1, FGFR2, and FGFR3, respectively); suppresses proliferation of cancer cells with wild-type FGFR3 overexpression (IC50s = 5, 30, 32, and 15 nM, for RT112, RT4, SW780, and JMSU1 cells, respectively); inhibited tumor growth in an RT112 bladder cancer xenograft mouse model overexpressing wild-type FGFR3 at 10-30 mg/kg
Catalog No. 50-291-7233 Supplier Cayman Chemical Supplier No. 3929210 MG
A pan FGFR inhibitor (IC50s = 0.9, 1.4, and 1 nM for FGFR1, FGFR2, and FGFR3, respectively); suppresses proliferation of cancer cells with wild-type FGFR3 overexpression (IC50s = 5, 30, 32, and 15 nM, for RT112, RT4, SW780, and JMSU1 cells, respectively); inhibited tumor growth in an RT112 bladder cancer xenograft mouse model overexpressing wild-type FGFR3 at 10-30 mg/kg