Learn More
New England Biolabs, Inc. BglI – 10000 units
SDP
Catalog No. 50812015
Encompass
A restriction endonuclease that recognizes the sequence GCCN_NNN^NGGC.
Catalog No. 50-812-015 Supplier New England Biolabs, Inc. Supplier No. R0143L
A restriction endonuclease that recognizes the sequence GCCN_NNN^NGGC.