Learn More
New England Biolabs, Inc. BglI – 2000 units
SDP
Catalog No. 50812016
Encompass
A restriction endonuclease that recognizes the sequence GCCN_NNN^NGGC.
Catalog No. 50-812-016 Supplier New England Biolabs, Inc. Supplier No. R0143S
A restriction endonuclease that recognizes the sequence GCCN_NNN^NGGC.