Learn More
New England Biolabs, Inc. BglII – 2000 units
SDP
Catalog No. 50812019
Encompass
A restriction endonuclease that recognizes the sequence A^GATC_T.
Catalog No. 50-812-019 Supplier New England Biolabs, Inc. Supplier No. R0144S
A restriction endonuclease that recognizes the sequence A^GATC_T.