Learn More
New England Biolabs, Inc. BglII – 10000 units
SDP
Supplier:  New England Biolabs, Inc. R0144M
Encompass
A restriction endonuclease that recognizes the sequence A^GATC_T.
Catalog No. 50-812-018