Learn More
Waters Corp Plunger Seal Kit, BGM, 4/pkg, for HPLC Systems
Catalog No. 502392904 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BGM Plunger Seal kit contains four each of the following: high pressure flanged seal, low-pressure seal, and a floating flanged seal-wash seal. The high- and low-pressure seals prevent pressurized solvent from leaking past the sapphire plunger.
  • Includes high pressure flanged seals
  • Includes low-pressure seals
  • Includes floating flanged seal-wash seals
  • Prevents pressurized solvent from leaking past the sapphire plunger
  • Suitable for pump head rebuilds
  • Designed for HPLC and purification systems
  • Compatible with auto purification instruments
  • Recommended for replacement as required
  • Component for the 2555 Quaternary Gradient Manager
Catalog No. 50-239-2904 Supplier Waters Corp Supplier No. 700001494
The BGM Plunger Seal kit contains four each of the following: high pressure flanged seal, low-pressure seal, and a floating flanged seal-wash seal. The high- and low-pressure seals prevent pressurized solvent from leaking past the sapphire plunger.
  • Includes high pressure flanged seals
  • Includes low-pressure seals
  • Includes floating flanged seal-wash seals
  • Prevents pressurized solvent from leaking past the sapphire plunger
  • Suitable for pump head rebuilds
  • Designed for HPLC and purification systems
  • Compatible with auto purification instruments
  • Recommended for replacement as required
  • Component for the 2555 Quaternary Gradient Manager