Learn More
Waters Corp Waters Performance Maintenance Kit, HPLC, Annual, for 2525/2545 BGM
Catalog No. 50818987 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This performance maintenance kit is designed to keep the 2525/2545 BGM instrument operating at peak performance on an annual basis. It contains all the necessary quality parts including sapphire plungers, high pressure plunger seals, a seal wash kit, low pressure seals, a check valve cartridge kit, solvent filters, a vent valve rotor seal, an inline filter frit, and a grease kit.
  • Performance maintenance type
  • HPLC system type
  • Annual replacement schedule
  • Includes sapphire plungers
  • Includes high pressure plunger seals
  • Includes seal wash kit
  • Includes low pressure seals
  • Includes check valve cartridge kit
  • Includes solvent filters
  • Includes vent valve rotor seal
  • Includes inline filter frit
  • Includes grease kit
  • One unit per package
Catalog No. 50-818-987 Supplier Waters Corp Supplier No. 201000130
Add to Cart
Add to Cart
Request a Quote
This performance maintenance kit is designed to keep the 2525/2545 BGM instrument operating at peak performance on an annual basis. It contains all the necessary quality parts including sapphire plungers, high pressure plunger seals, a seal wash kit, low pressure seals, a check valve cartridge kit, solvent filters, a vent valve rotor seal, an inline filter frit, and a grease kit.
  • Performance maintenance type
  • HPLC system type
  • Annual replacement schedule
  • Includes sapphire plungers
  • Includes high pressure plunger seals
  • Includes seal wash kit
  • Includes low pressure seals
  • Includes check valve cartridge kit
  • Includes solvent filters
  • Includes vent valve rotor seal
  • Includes inline filter frit
  • Includes grease kit
  • One unit per package