Learn More
Aviva Systems Biology Corp BGN Antibody - middle region (ARP56333_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56333_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BGN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BGN. Alternate names: DSPG1, PG-S1, PGI, SLRR1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61814