Learn More
Selleck Chemical LLC BGP-15 2HCl
SDP
Catalog No. 5000000820
Encompass_Preferred
BGP-15 is a nicotinic amidoxime derivative with PARP inhibitory activity It has been demonstrated that BGP-15 protects against ischemia-reperfusion injury
Catalog No. 50-000-00820 Supplier Selleck Chemical LLC Supplier No. S837025MG
Add to Cart
Add to Cart
BGP-15 is a nicotinic amidoxime derivative with PARP inhibitory activity It has been demonstrated that BGP-15 protects against ischemia-reperfusion injury