Learn More
Medchemexpress LLC HY-100828 5mg Medchemexpress, BGP-15 CAS:66611-37-8 Purity:>98%
SDP
Catalog No. 501931964
Encompass_Preferred
Medchemexpress, HY-100828 5mg BGP-15 CAS:66611-37-8 BGP-15 is a PARP inhibitor, with an IC 50 and a K i of 120 and 57 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1964 Supplier Medchemexpress LLC Supplier No. HY1008285MG
Medchemexpress, HY-100828 5mg BGP-15 CAS:66611-37-8 BGP-15 is a PARP inhibitor, with an IC 50 and a K i of 120 and 57 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.