Learn More
TARGETMOL CHEMICALS INC BGT226 5MG
SDP
Catalog No. 502576609
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. BGT226 (NVP-BGT226) is a novel dual PI3K / mTOR inhibitor it acts on PI3K α/β/γ with IC50 of 4 nM / 63 nM / 38 nM respectively. purity: 99%
Catalog No. 50-257-6609 Supplier TARGETMOL CHEMICALS INC Supplier No. T6072L5MG
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. BGT226 (NVP-BGT226) is a novel dual PI3K / mTOR inhibitor it acts on PI3K α/β/γ with IC50 of 4 nM / 63 nM / 38 nM respectively. purity: 99%