Learn More
Medchemexpress LLC BGT226 (maleate) | 1245537-68-1 | MFCD22124895 | 99.9% | 650.60 g/mol | C32H29F3N6O6 | 5 MG
SDP
Catalog No. 5000262390
Encompass_Preferred
BGT226 maleate is a small-molecule PI3K/mTOR dual inhibitor used for research into PI3K signaling and oncology. It shows low-nanomolar potency against PI3K isoforms and mTOR and is used in cell-based and preclinical studies to evaluate antiproliferative and apoptotic effects.
  • Inhibits PI3Kα, PI3Kβ, PI3Kγ, and mTOR.
  • Exhibits low nanomolar potency in enzymatic assays.
  • Demonstrates growth-inhibitory activity in cancer cell models.
  • Supplied as the maleate salt for stability.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple small pack sizes for research use.
Catalog No. 50-002-62390 Supplier Medchemexpress LLC Supplier No. HY133345MG
BGT226 maleate is a small-molecule PI3K/mTOR dual inhibitor used for research into PI3K signaling and oncology. It shows low-nanomolar potency against PI3K isoforms and mTOR and is used in cell-based and preclinical studies to evaluate antiproliferative and apoptotic effects.
  • Inhibits PI3Kα, PI3Kβ, PI3Kγ, and mTOR.
  • Exhibits low nanomolar potency in enzymatic assays.
  • Demonstrates growth-inhibitory activity in cancer cell models.
  • Supplied as the maleate salt for stability.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple small pack sizes for research use.