Learn More
Medchemexpress LLC Bgt226 maleate | 1245537-68-1 | MFCD22124895 | 99.9% | C32H29F3N6O6 | 10MG
SDP
Catalog No. 5000259479
Encompass_Preferred
BGT226 maleate is a research-grade small-molecule inhibitor of class I PI3K isoforms and mTOR, used to probe PI3K/mTOR signaling in cellular and biochemical studies. The compound is supplied as a high-purity solid salt for laboratory research use only and is provided with recommended storage conditions and analytical quality data.
Catalog No. 50-002-59479 Supplier Medchemexpress LLC Supplier No. HY1333410MG
BGT226 maleate is a research-grade small-molecule inhibitor of class I PI3K isoforms and mTOR, used to probe PI3K/mTOR signaling in cellular and biochemical studies. The compound is supplied as a high-purity solid salt for laboratory research use only and is provided with recommended storage conditions and analytical quality data.