Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BH3 (hydrochloride) | 98.03% | 2677.45 | 5 MG
SDP

Catalog No. 5000422698
Encompass_Preferred

BH3 hydrochloride is a BBB penetrated peptide that provokes apoptosis either by direct activation of pro-apoptotic Bax/Bak or by neutralizing anti-apoptotic Bcl-2 proteins (Bcl-2, Bcl-XL, Bcl-w, Mcl-1, and A-1) via their BH3 domain.

  • BBB penetrated peptide
  • Provokes apoptosis by direct activation of pro-apoptotic Bax/Bak
  • Neutralizes anti-apoptotic Bcl-2 proteins
  • Targets Bcl-2, Bcl-XL, Bcl-w, Mcl-1, and A-1 via their BH3 domain
  • Purity: 98.03%
  • Molecular weight: 2677.45
  • Solid, white to off-white appearance
  • Soluble in water at ≥ 33.33 mg/mL
  • For research use only

Catalog No. 50-004-22698 Supplier Medchemexpress LLC Supplier No. HYP23435MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BH3 hydrochloride is a BBB penetrated peptide that provokes apoptosis either by direct activation of pro-apoptotic Bax/Bak or by neutralizing anti-apoptotic Bcl-2 proteins (Bcl-2, Bcl-XL, Bcl-w, Mcl-1, and A-1) via their BH3 domain.

  • BBB penetrated peptide
  • Provokes apoptosis by direct activation of pro-apoptotic Bax/Bak
  • Neutralizes anti-apoptotic Bcl-2 proteins
  • Targets Bcl-2, Bcl-XL, Bcl-w, Mcl-1, and A-1 via their BH3 domain
  • Purity: 98.03%
  • Molecular weight: 2677.45
  • Solid, white to off-white appearance
  • Soluble in water at ≥ 33.33 mg/mL
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.