Learn More
Medchemexpress LLC Bh3i-1 | 300817-68-9 | MFCD03453544 | 98.0% | 400.31 | C15H14BrNO3S2 | 100 MG
SDP
Catalog No. 5000213532
Encompass_Preferred
BH3I-1 is a small-molecule antagonist of the Bcl-2 family that disrupts BH3-domain interactions. Reported activities include inhibition of Bak BH3 binding to Bcl-xL (Ki 2.4 ±0.2 μM), binding to the p53/MDM2 pair (Kd 5.3 μM), and NMR evidence of targeting the BH3-binding pocket (Ki 7.8 ±0.9 μM). Supplied at ~98% purity for research use.
  • Inhibits Bak BH3 peptide binding to Bcl-xL (Ki ≈2.4 μM).
  • Binds p53/MDM2 with a reported Kd of 5.3 μM.
  • Targets the BH3-binding pocket of Bcl-xL (NMR Ki ≈7.8 μM).
  • High purity suitable for biochemical and cellular studies.
  • Available in small-scale quantities for laboratory use.
Catalog No. 50-002-13532 Supplier Medchemexpress LLC Supplier No. HY100383100MG
BH3I-1 is a small-molecule antagonist of the Bcl-2 family that disrupts BH3-domain interactions. Reported activities include inhibition of Bak BH3 binding to Bcl-xL (Ki 2.4 ±0.2 μM), binding to the p53/MDM2 pair (Kd 5.3 μM), and NMR evidence of targeting the BH3-binding pocket (Ki 7.8 ±0.9 μM). Supplied at ~98% purity for research use.
  • Inhibits Bak BH3 peptide binding to Bcl-xL (Ki ≈2.4 μM).
  • Binds p53/MDM2 with a reported Kd of 5.3 μM.
  • Targets the BH3-binding pocket of Bcl-xL (NMR Ki ≈7.8 μM).
  • High purity suitable for biochemical and cellular studies.
  • Available in small-scale quantities for laboratory use.