Learn More
Azzota Corp BHA350T UC 15L 53kHz 350W 33x3
Catalog No. 5000887250
Encompass_Preferred
Model BHA350T Adjustable Power Cleaner Benchtop 53 kHz 15L Temp range 20-60C 350W 40%-100% Power adjustable internal size 33x30x15cm
Catalog No. 50-008-87250 Supplier Azzota Corp Supplier No. BHA350T
Model BHA350T Adjustable Power Cleaner Benchtop 53 kHz 15L Temp range 20-60C 350W 40%-100% Power adjustable internal size 33x30x15cm