Learn More
Cayman Chemical BHC 100mg
SDP
Catalog No. 502936200
Encompass_Preferred
An HIV maturation inhibitor (IC90 6.4 nM) active against a variety of HIV isolates
Catalog No. 50-293-6200 Supplier Cayman Chemical Supplier No. 24134 100MG
An HIV maturation inhibitor (IC90 6.4 nM) active against a variety of HIV isolates