Learn More
Cayman Chemical BHC 50mg
SDP
Catalog No. 502935567
Encompass_Preferred
A selective allosteric orally bioavailable pan-Akt inhibitor (IC50s 5.0 4.5 and 16 nM for Akt1 Akt2 and Akt3 respectively)
Catalog No. 50-293-5567 Supplier Cayman Chemical Supplier No. 24134 50MG
A selective allosteric orally bioavailable pan-Akt inhibitor (IC50s 5.0 4.5 and 16 nM for Akt1 Akt2 and Akt3 respectively)