Learn More
Azzota Corp BHF150 UC 4.5L 53kHz 150W LCD
Catalog No. 5000887243
Encompass_Preferred
Model BHF150 Ultrasonic Cleaner Benchtop 53 kHz Frequency 4 5L Capacity 150 W 1-99 Min Timer LCD Display
Catalog No. 50-008-87243 Supplier Azzota Corp Supplier No. BHF150
Model BHF150 Ultrasonic Cleaner Benchtop 53 kHz Frequency 4 5L Capacity 150 W 1-99 Min Timer LCD Display