Learn More
AdipoGen BHHCT
SDP
Catalog No. 502560784
Encompass_Preferred
Chemical. CAS 200862-70-0. Formula C30H15ClF14O6S. MW 804.93. Synthetic. Sensitive label for time-resolved fluoroimmunoassay via the europium chelate. Ligand for TRF-complexes with Eu3 used for preparing Silica-Nanoparticles. This label is characterized by very large Stokes shift, broad exitation and narrow emission bands.
Catalog No. 50-256-0784 Supplier AdipoGen Supplier No. CDXH0041G001
Chemical. CAS 200862-70-0. Formula C30H15ClF14O6S. MW 804.93. Synthetic. Sensitive label for time-resolved fluoroimmunoassay via the europium chelate. Ligand for TRF-complexes with Eu3 used for preparing Silica-Nanoparticles. This label is characterized by very large Stokes shift, broad exitation and narrow emission bands.