Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp BHI Agar [Brain Heart Infusion Agar], 500 Grams
SDP

Catalog No. 50488529
Encompass_Preferred

Recommended for the cultivation of a wide variety of microorganisms including many types of pathogens. BHI Agar is currently recommended as a general medium for aerobic bacteria and for the primary recovery of fungi from clinical specimens.

Dissolve 50 grams in 1L distilled water. Sterilize by autoclaving 15 minutes.

Ingredients:

Brain Heart Infusion from solids: 17.5 g/l
Pancreatic Digest of Gelatin: 10.0 g/l
Dextrose: 2.0 g/l
Sodium Chloride: 5.0 g/l
Disodium Phosphate: 2.5 g/l
Agar: 13.5 g/l

Catalog No. 50-488-529 Supplier Research Products International Corp Supplier No. B11500500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recommended for the cultivation of a wide variety of microorganisms including many types of pathogens. BHI Agar is currently recommended as a general medium for aerobic bacteria and for the primary recovery of fungi from clinical specimens.

Dissolve 50 grams in 1L distilled water. Sterilize by autoclaving 15 minutes.

Ingredients:

Brain Heart Infusion from solids: 17.5 g/l
Pancreatic Digest of Gelatin: 10.0 g/l
Dextrose: 2.0 g/l
Sodium Chloride: 5.0 g/l
Disodium Phosphate: 2.5 g/l
Agar: 13.5 g/l

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.