Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ BHI Agar Plate
SDP

Catalog No. 23001809 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-809 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-809 Supplier Hardy Diagnostics™ Supplier No. W163
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Brain Heart Infusion (BHI) Agar is a general purpose nutrient medium recommended for the cultivation and isolation of a variety of microorganisms, including bacteria, yeasts, and molds.

  • Organic nitrogen, carbon, sulfur, vitamins and trace substances are provided by the peptones and infusion
  • Dextrose provides the carbohydrate source for fermentative microorganisms
  • Disodium phosphate is added to the medium in order to maintain an optimal pH
  • A variety of supplements has been added to further enhance the recovery of fastidious microorganisms

Specifications

Appearance Slightly Opalescent
Color Light Amber
Content And Storage 2°C to 8°C
Detects Bacteria, Yeasts, Molds
Form Prepared Medium
Format Plate
Media Type Agar
Product Type Agar Plate
Test Type Microbial Cultivation
Fill Volume 60 mL
For Use With (Application) Cannabis QA Testing, Clinical, Cosmetics, Environmental
Incubation Conditions 35°C
Incubation Time 24 to 48 hr.
No. per Pack 5
Quantity 5 Ea.
Dimensions (Dia. x H) 100 x 25 mm
Height (Metric) 25 mm
Diameter (Metric) 100 mm
Species Escherichia coli, Staphylococcus species, Staphylococcus aureus
Protein Source Beef, Pork, Casein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.