Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ BHI Agar with Vancomycin
SDP

Catalog No. 23001794 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Species:
Enterococcus faecalis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-001-794 Enterococcus faecalis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-794 Supplier Hardy Diagnostics™ Supplier No. G14
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Brain Heart Infusion Agar contains vancomycin for the detection and isolation of vancomycin resistant Enterococcus spp.

  • 6 μg/mL of vancomycin
  • If an organism does not grow on this medium, it is considered susceptible; if the organism does grow, it is considered resistant to vancomycin

Specifications

Appearance Translucent
Color Light Amber
Content And Storage 2°C to 8°C
Additive Vancomycin
Detects Vancomycin-resistant Enterococcus spp.
Form Prepared Medium
Format Plate
Media Type BHI Agar
Product Type Agar Plate
Sterility Sterile
Target Organism Class Gram-positive Cocci
Test Type Antibiotic Susceptibility Sensitivity
Fill Volume 18 mL
Formulation Beef Heart Infusion - 250.0gm, Calf Brain Infusion - 200.0gm, Proteose Peptone - 10.0gm, Sodium Chloride - 5.0gm, Disodium Phosphate - 2.5gm, Dextrose - 2.0gm, Vancomycin - 6.0mg, Agar - 15.0gm
For Use With (Application) Antibiotic Susceptibility Testing
Incubation Conditions 35°C
Incubation Time 24 hr.
No. per Pack 10
Packaging Type Plates, Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Enterococcus faecalis
Protein Source Beef Heart Infusion, Calf Brain Infusion, Proteose Peptone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.