Learn More
Alkali Scientific BHI Broth [Brain Heart Infusion Broth], 1 Kg
SDP
Catalog No. 5000660775
Encompass_Preferred
Brain Heart Infusion (BHI) Broth, 1 Kg, is a nutrient-rich medium commonly used for growing fastidious microorganisms, including bacteria, fungi, and yeast. The powder formulation contains essential nutrients like amino acids, vitamins, and growth factors that support the propagation and isolation of a wide range of microorganisms. BHI Broth is crucial for microbial cultivation in clinical diagnostics, pharmaceutical microbiology, and food safety testing. It is often used to grow pathogenic microorganisms, enabling the study of their behavior and characteristics under controlled laboratory conditions. This 1 Kg size is ideal for large-scale preparation of media in high-throughput laboratories.
Catalog No. 50-006-60775 Supplier Alkali Scientific Supplier No. BHI002CP
Brain Heart Infusion (BHI) Broth, 1 Kg, is a nutrient-rich medium commonly used for growing fastidious microorganisms, including bacteria, fungi, and yeast. The powder formulation contains essential nutrients like amino acids, vitamins, and growth factors that support the propagation and isolation of a wide range of microorganisms. BHI Broth is crucial for microbial cultivation in clinical diagnostics, pharmaceutical microbiology, and food safety testing. It is often used to grow pathogenic microorganisms, enabling the study of their behavior and characteristics under controlled laboratory conditions. This 1 Kg size is ideal for large-scale preparation of media in high-throughput laboratories.