Learn More
Alkali Scientific BHI Broth [Brain Heart Infusion Broth], 5 Kg
SDP
Catalog No. 5000661668
Encompass_Preferred
Brain Heart Infusion (BHI) Broth, 5 Kg, is a nutrient-rich medium used for the growth of fastidious bacteria, fungi, and yeast. This large 5 Kg pack size is ideal for high-throughput laboratories that require a large volume of media for microbial cultivation. The medium contains essential nutrients such as amino acids, vitamins, and growth factors that support the propagation and isolation of a wide range of microorganisms. BHI Broth is crucial for microbiological applications in clinical diagnostics, pharmaceutical testing, and food safety testing. It is widely used for cultivating pathogens and studying microbial behaviors under controlled conditions.
Catalog No. 50-006-61668 Supplier Alkali Scientific Supplier No. BHI004CP
Brain Heart Infusion (BHI) Broth, 5 Kg, is a nutrient-rich medium used for the growth of fastidious bacteria, fungi, and yeast. This large 5 Kg pack size is ideal for high-throughput laboratories that require a large volume of media for microbial cultivation. The medium contains essential nutrients such as amino acids, vitamins, and growth factors that support the propagation and isolation of a wide range of microorganisms. BHI Broth is crucial for microbiological applications in clinical diagnostics, pharmaceutical testing, and food safety testing. It is widely used for cultivating pathogens and studying microbial behaviors under controlled conditions.