Learn More
HIMEDIA BHI Broth, HiVeg, MV210, Microbiology Culture Media, Dehydrated Culture Media, Broth, Animal-Component-Free, General Purpose Enrichment, Powder, 500 g
Catalog No. NC4013776
Encompass_Preferred
BHI Broth, HiVeg is an animal-component-free brain heart infusion broth for the general-purpose propagation of pathogenic cocci and other fastidious organisms in blood culture work and allied pathological investigations. The plant-based HiVeg infusions and peptone, with dextrose and buffering phosphates at final pH 7.4, deliver the rich nutrition expected of BHI while removing animal-origin components. Ideal as an enrichment and pre-culture broth for a broad organism range. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013776 Supplier HIMEDIA Supplier No. MV210500G
BHI Broth, HiVeg is an animal-component-free brain heart infusion broth for the general-purpose propagation of pathogenic cocci and other fastidious organisms in blood culture work and allied pathological investigations. The plant-based HiVeg infusions and peptone, with dextrose and buffering phosphates at final pH 7.4, deliver the rich nutrition expected of BHI while removing animal-origin components. Ideal as an enrichment and pre-culture broth for a broad organism range. Dehydrated powder, store in a cool dry place. Packaged as 500 g.