Learn More
AG Scientific Inc BHI Broth, Powder, 500 G
Catalog No. 502237061
Encompass_Preferred
Brain Heart Infusion Broth (BHI broth) is a useful growth medium for many types of microorganisms, including Streptococci, Pneumococci, and Meningococci just to name a few. BHI growth medium is commonly used in food safety, water safety, and antibiotic sensitivity tests. It is particularly useful for cultivating rare microbe species. Growth of anaerobic organisms can be enhanced by adding 0.1% agar to the BHI broth.Solubility: WaterStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7061 Supplier AG Scientific Inc Supplier No. B2587500GM
Brain Heart Infusion Broth (BHI broth) is a useful growth medium for many types of microorganisms, including Streptococci, Pneumococci, and Meningococci just to name a few. BHI growth medium is commonly used in food safety, water safety, and antibiotic sensitivity tests. It is particularly useful for cultivating rare microbe species. Growth of anaerobic organisms can be enhanced by adding 0.1% agar to the BHI broth.Solubility: WaterStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.