Learn More
Eurofins Discoverx PathHunter BHK-21 rPTGER4 β-Arrestin Cell Line, 2 vials. (93-0908C16)
Supplier:  Eurofins Discoverx 930908C16
Encompass
The PathHunter rPTGER4 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. [ligand]) based activation of rPTGER4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rPTGER4 and EA-tagged β-Arrestin-2. Activation of rPTGER4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1545
Add to Cart
Request a Quote