Learn More
ATCC BHK-21 (C-13); Kidney; Syrian golden hamster (Mesocricetus auratus)
Catalog No. 502381685
Encompass
BHK-21 [13] fibroblasts are isolated from the kidney of an uninfected golden hamster. The World Organization for Animal Health (OIE) s these cells for routine diagnosis of rabies. This cell line has been used as a host for transformation with expression vectors containing selectable and amplifiable marker DNAs.
Catalog No. 50-238-1685 Supplier ATCC Supplier No. CCL10
BHK-21 [13] fibroblasts are isolated from the kidney of an uninfected golden hamster. The World Organization for Animal Health (OIE) s these cells for routine diagnosis of rabies. This cell line has been used as a host for transformation with expression vectors containing selectable and amplifiable marker DNAs.