Learn More
Aviva Systems Biology Corp BHLHB2 Antibody - middle region (ARP34006_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34006_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BHLHB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BHLHB2. Alternate names: DEC1, SHARP-2, STRA13, Stra14, HLHB2, BHLHB2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-55218