Learn More
Aviva Systems Biology Corp BHLHB5 Antibody - N-terminal region (ARP32430_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32430_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against BHLHB5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BHLHB5. Alternate names: Beta3, BHLHB5, CAGL85, TNRC20ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54585