Learn More
Aviva Systems Biology Corp Bhlhe41 Antibody - N-terminal region (ARP39651_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39651_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Bhlhe41. It was validated on Western Blot. . Alternate names: Bhlhb3, Dec2, SHARP-1, Sharp1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-284-820