Learn More
Apexbio Technology LLC BHQ 88-58-4 100mg
SDP
Catalog No. 502240408
Encompass_Preferred
BHQ (2 5-di-tert-butyl-1 4-benzohydroquinone CAS 88-58-4) is a selective inhibitor of the endoplasmic reticulum Ca -ATPase which is responsible for transporting calcium from the cytosol into the sarcoplasmic or endoplasmic reticulum In rat basophilic leukemia cells 10 M BHQ inhibits inward rectifier potassium currents and may induce cell depolarization and affect calcium influx In vascular smooth muscle studies BHQ modulates calcium-induced contractions in a K -dependent manner and inhibits L-type calcium currents (IC 66 7 M) via superoxide anion generation In MDCK cells BHQ increases intracellular calcium in a dose-dependent manner (EC 40 M) by depleting ER calcium stores BHQ is widely used to investigate calcium signaling and ER function
This item is not returnable. View return policy
Catalog No. 50-224-0408 Supplier Apexbio Technology LLC Supplier No. B6648100
BHQ (2 5-di-tert-butyl-1 4-benzohydroquinone CAS 88-58-4) is a selective inhibitor of the endoplasmic reticulum Ca -ATPase which is responsible for transporting calcium from the cytosol into the sarcoplasmic or endoplasmic reticulum In rat basophilic leukemia cells 10 M BHQ inhibits inward rectifier potassium currents and may induce cell depolarization and affect calcium influx In vascular smooth muscle studies BHQ modulates calcium-induced contractions in a K -dependent manner and inhibits L-type calcium currents (IC 66 7 M) via superoxide anion generation In MDCK cells BHQ increases intracellular calcium in a dose-dependent manner (EC 40 M) by depleting ER calcium stores BHQ is widely used to investigate calcium signaling and ER function