Learn More
Medchemexpress LLC BHQ-2 NHS | 916753-62-3 | 97.7% | 603.58 | 5 MG
SDP
Catalog No. 5000404727
Encompass_Preferred
BHQ-2 NHS is a dark quencher lacking native emission, attributed to its polyaromatic-azo backbone and a terminal NHS ester. It features a broad and intense quenching range from 560-670 nm, making it suitable for fluorescence resonance energy transfer (FRET) applications when used with orange to far-red emitting dyes. The NHS ester allows for labeling of primary amines (-NH2) on proteins, amine-modified oligonucleotides, and other amine-containing molecules.
  • Dark quencher with no native emission.
  • Polyaromatic-azo backbone and a terminal NHS ester.
  • Wide and intense quenching range from 560-670 nm.
  • Useful as an acceptor in FRET applications.
  • Can label primary amines (-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules.
  • For research use only.
Catalog No. 50-004-04727 Supplier Medchemexpress LLC Supplier No. HY429845MG
BHQ-2 NHS is a dark quencher lacking native emission, attributed to its polyaromatic-azo backbone and a terminal NHS ester. It features a broad and intense quenching range from 560-670 nm, making it suitable for fluorescence resonance energy transfer (FRET) applications when used with orange to far-red emitting dyes. The NHS ester allows for labeling of primary amines (-NH2) on proteins, amine-modified oligonucleotides, and other amine-containing molecules.
  • Dark quencher with no native emission.
  • Polyaromatic-azo backbone and a terminal NHS ester.
  • Wide and intense quenching range from 560-670 nm.
  • Useful as an acceptor in FRET applications.
  • Can label primary amines (-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules.
  • For research use only.